CAS RN 73279-04-6
CAS RN 73279-04-6
IUPAC Name:
3-[(piperidin-1-yl)methyl]phenol
Synonyms:
3-(piperidin-1-ylmethyl)phenol
Molecular Weight (g/mol):
191.27
Molecular Formula:
C12H17NO
InChi Key:
ORGBERFQYFWYGX-UHFFFAOYSA-N
SMILES:
OC1=CC=CC(CN2CCCCC2)=C1
1
–
1
of
1
results
1
1-(3-Hydroxybenzyl)piperidine 98.0+%, TCI America™
CAS: 73279-04-6 Molecular Formula: C12H17NO Molecular Weight (g/mol): 191.274 MDL Number: MFCD00800250 InChI Key: ORGBERFQYFWYGX-UHFFFAOYSA-N Synonym: 3-1-piperidinylmethyl phenol,3-piperidin-1-ylmethyl phenol,3-1-piperidinomethyl phenol,1-3-hydroxybenzyl piperidine,3-piperidinomethyl phenol,3-1-piperidylmethyl phenol,phenol, 3-1-piperidinylmethyl,1-3-hydroxyphenylmethyl piperidine,pubchem6815,3-piperidinomethylphenol PubChem CID: 826974 IUPAC Name: 3-(piperidin-1-ylmethyl)phenol SMILES: C1CCN(CC1)CC2=CC(=CC=C2)O