missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(3-Fluorobenzyl)piperazine 98.0+%, TCI America™
$61.51 - $212.51
Chemical Identifiers
| CAS | 55513-19-4 |
|---|---|
| Molecular Formula | C11H15FN2 |
| Molecular Weight (g/mol) | 194.253 |
| MDL Number | MFCD02242870 |
| InChI Key | ITHBJSRWFNLKIH-UHFFFAOYSA-N |
| Synonym | 1-3-fluorobenzyl piperazine, 1-3-fluorophenyl methyl piperazine, 1-3-fluoro-benzyl-piperazine, 3-fluorophenyl methyl piperazine, 1-3-fluorobenz-yl piperazine, piperazine, 1-3-fluorophenyl methyl, 1-3-fluorophenyl methyl piperazine trifluoroacetate |
| PubChem CID | 903813 |
| IUPAC Name | 1-[(3-fluorophenyl)methyl]piperazine |
| SMILES | C1CN(CCN1)CC2=CC(=CC=C2)F |
Chemical Identifiers
| 55513-19-4 | |
| 194.253 | |
| ITHBJSRWFNLKIH-UHFFFAOYSA-N | |
| 903813 | |
| C1CN(CCN1)CC2=CC(=CC=C2)F |
| C11H15FN2 | |
| MFCD02242870 | |
| 1-3-fluorobenzyl piperazine, 1-3-fluorophenyl methyl piperazine, 1-3-fluoro-benzyl-piperazine, 3-fluorophenyl methyl piperazine, 1-3-fluorobenz-yl piperazine, piperazine, 1-3-fluorophenyl methyl, 1-3-fluorophenyl methyl piperazine trifluoroacetate | |
| 1-[(3-fluorophenyl)methyl]piperazine |
Specifications
| 55513-19-4 | |
| 115°C | |
| MFCD02242870 | |
| 1-3-fluorobenzyl piperazine, 1-3-fluorophenyl methyl piperazine, 1-3-fluoro-benzyl-piperazine, 3-fluorophenyl methyl piperazine, 1-3-fluorobenz-yl piperazine, piperazine, 1-3-fluorophenyl methyl, 1-3-fluorophenyl methyl piperazine trifluoroacetate | |
| C1CN(CCN1)CC2=CC(=CC=C2)F | |
| 194.253 | |
| 194.25 | |
| Liquid |
| Yellow | |
| C11H15FN2 | |
| 1 g | |
| ITHBJSRWFNLKIH-UHFFFAOYSA-N | |
| 1-[(3-fluorophenyl)methyl]piperazine | |
| 903813 | |
| ≥98.0% (GC,T) | |
| 1-(3-Fluorobenzyl)piperazine |
Safety and Handling
TSCA : No