missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Fluorobenzyl)piperazine 98.0+%, TCI America™

Catalog No. F10925G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10925G 5 g
F10921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10925G Supplier TCI America Supplier No. F10925G

Chemical Identifiers

CAS 55513-19-4
Molecular Formula C11H15FN2
Molecular Weight (g/mol) 194.253
MDL Number MFCD02242870
InChI Key ITHBJSRWFNLKIH-UHFFFAOYSA-N
Synonym 1-3-fluorobenzyl piperazine, 1-3-fluorophenyl methyl piperazine, 1-3-fluoro-benzyl-piperazine, 3-fluorophenyl methyl piperazine, 1-3-fluorobenz-yl piperazine, piperazine, 1-3-fluorophenyl methyl, 1-3-fluorophenyl methyl piperazine trifluoroacetate
PubChem CID 903813
IUPAC Name 1-[(3-fluorophenyl)methyl]piperazine
SMILES C1CN(CCN1)CC2=CC(=CC=C2)F

Specifications

Color Yellow
Boiling Point 115°C
Quantity 5 g
Formula Weight 194.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(3-Fluorobenzyl)piperazine