missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Fluorobenzyl)piperazine, 97%

Catalog No. p-7052683
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH5529503 1 g
AAH5529506 5 g
2 options

Catalog No. AAH5529503

Supplier: Thermo Scientific Chemicals H5529503

Only null left

CAS: 55513-19-4 | C11H15FN2 | 194.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55513-19-4
Molecular Formula C11H15FN2
Molecular Weight (g/mol) 194.253
MDL Number MFCD02242870
InChI Key ITHBJSRWFNLKIH-UHFFFAOYSA-N
Synonym 1-3-fluorobenzyl piperazine, 1-3-fluorophenyl methyl piperazine, 1-3-fluoro-benzyl-piperazine, 3-fluorophenyl methyl piperazine, 1-3-fluorobenz-yl piperazine, piperazine, 1-3-fluorophenyl methyl, 1-3-fluorophenyl methyl piperazine trifluoroacetate
PubChem CID 903813
IUPAC Name 1-[(3-fluorophenyl)methyl]piperazine
SMILES C1CN(CCN1)CC2=CC(=CC=C2)F

Specifications

Color Colorless to Orange
Density 1.106 g/mL
Boiling Point 281°C
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 1 g
Formula Weight 194.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(3-Fluorobenzyl)piperazine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only