missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dithiole-2-thione 97.0+%, TCI America™
$80.16 - $402.63
Chemical Identifiers
| CAS | 930-35-8 |
|---|---|
| Molecular Formula | C3H2S3 |
| Molecular Weight (g/mol) | 134.23 |
| MDL Number | MFCD00014536 |
| InChI Key | WYKJWNVWJOKVQP-UHFFFAOYSA-N |
| PubChem CID | 70254 |
| IUPAC Name | 2H-1,3-dithiole-2-thione |
| SMILES | S=C1SC=CS1 |
Chemical Identifiers
| 930-35-8 | |
| 134.23 | |
| WYKJWNVWJOKVQP-UHFFFAOYSA-N | |
| 2H-1,3-dithiole-2-thione |
| C3H2S3 | |
| MFCD00014536 | |
| 70254 | |
| S=C1SC=CS1 |
Specifications
| 930-35-8 | |
| C3H2S3 | |
| 1 g | |
| S=C1SC=CS1 | |
| 134.23 | |
| 134.23 | |
| Crystalline Powder |
| 49°C | |
| MFCD00014536 | |
| WYKJWNVWJOKVQP-UHFFFAOYSA-N | |
| 2H-1,3-dithiole-2-thione | |
| 70254 | |
| ≥97.0% (GC) | |
| 1,3-Dithiole-2-thione |
Safety and Handling
TSCA : No