accessibility menu, dialog, popup

UserName

SSS

CAS RN 930-35-8

IUPAC Name: 2H-1,3-dithiole-2-thione
Synonyms: 1,3-dithiole-2-thione
Molecular Weight (g/mol): 134.23
Molecular Formula: C3H2S3
InChi Key: WYKJWNVWJOKVQP-UHFFFAOYSA-N
SMILES: S=C1SC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results