missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| 1,3-Dithiane | |
| 53°C | |
| C4H8S2 | |
| 5 g | |
| WQADWIOXOXRPLN-UHFFFAOYSA-N | |
| 1,3-dithiane | |
| 10451 | |
| 120.23 | |
| Crystalline Powder |
| 505-23-7 | |
| 208°C | |
| MFCD00006654 | |
| 1,3-dithiacyclohexane, m-dithiane, dithiane-1,3, 1,3-dithian, dithiane-1,3 french, 1,3 dithiane, ccris 6777, m-dithiane, 8ci, 1,3 dithiocyclohexane, acmc-209knk | |
| C1CSCSC1 | |
| 120.228 | |
| CHEBI:47845 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 505-23-7 | |
| 120.228 | |
| WQADWIOXOXRPLN-UHFFFAOYSA-N | |
| 10451 | |
| 1,3-dithiane |
| C4H8S2 | |
| MFCD00006654 | |
| 1,3-dithiacyclohexane, m-dithiane, dithiane-1,3, 1,3-dithian, dithiane-1,3 french, 1,3 dithiane, ccris 6777, m-dithiane, 8ci, 1,3 dithiocyclohexane, acmc-209knk | |
| CHEBI:47845 | |
| C1CSCSC1 |
Safety and Handling
RTECSNumber : JO5070000
TSCA : No