accessibility menu, dialog, popup

UserName

SS

CAS RN 505-23-7

IUPAC Name: 1,3-dithiane
Synonyms: 1,3-dithiane dithiane-1,3
Molecular Weight (g/mol): 120.23
Molecular Formula: C4H8S2
InChi Key: WQADWIOXOXRPLN-UHFFFAOYSA-N
SMILES: C1CSCSC1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)