missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dithiane, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 169510250

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC169510250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

505-23-7
120.23
WQADWIOXOXRPLN-UHFFFAOYSA-N
10451
1,3-dithiane
C4H8S2
MFCD00006654
1,3-dithiacyclohexane, m-dithiane, dithiane-1,3, 1,3-dithian, dithiane-1,3 french, 1,3 dithiane, ccris 6777, m-dithiane, 8ci, 1,3 dithiocyclohexane, acmc-209knk
CHEBI:47845
C1CSCSC1

Specifications

1, 3-Dithiane
96.0
90g/mL
Authentic
Glass bottle
25g
02,182; 03,135; 04,216; 05,287; 06,248; 07,142; 14,157; 16,161; 01,208; 02,108; 12,158; 15,112
WQADWIOXOXRPLN-UHFFFAOYSA-N
1,3-dithiane
10451
120.23
505-23-7
100.0
90°C
97%
C4H8S2
MFCD00006654
1,3-dithiacyclohexane, m-dithiane, dithiane-1,3, 1,3-dithian, dithiane-1,3 french, 1,3 dithiane, ccris 6777, m-dithiane, 8ci, 1,3 dithiocyclohexane, acmc-209knk
C1CSCSC1
120.23
CHEBI:47845
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-006-7

SDS
Documents

Documents

Promotions

Promotions