missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dithiane, 97%

Catalog No. AC169510050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169510050 5 g Glass bottle
AC169510250 25 g Glass bottle
AC169511000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC169510050 Supplier Thermo Scientific Chemicals Supplier No. 169510050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 505-23-7 | C4H8S2 | 120.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 505-23-7
Molecular Formula C4H8S2
Molecular Weight (g/mol) 120.23
MDL Number MFCD00006654
InChI Key WQADWIOXOXRPLN-UHFFFAOYSA-N
Synonym 1,3-dithiacyclohexane, m-dithiane, dithiane-1,3, 1,3-dithian, dithiane-1,3 french, 1,3 dithiane, ccris 6777, m-dithiane, 8ci, 1,3 dithiocyclohexane, acmc-209knk
PubChem CID 10451
ChEBI CHEBI:47845
IUPAC Name 1,3-dithiane
SMILES C1CSCSC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 49°C to 54°C
Color White to Beige
Density 90g/mL
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Fieser 02,182; 03,135; 04,216; 05,287; 06,248; 07,142; 14,157; 16,161; 01,208; 02,108; 12,158; 15,112
Formula Weight 120.23
Percent Purity 97%
Physical Form Crystals, Needles or Powder
Chemical Name or Material 1, 3-Dithiane
Show More Show Less
EINECSNumber 208-006-7

RUO – Research Use Only