missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylurea 98.0+%, TCI America™

Catalog No. C003125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C003125G 25 g
C0031100G 100 g
C0031500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C003125G Supplier TCI America Supplier No. C003125G

Chemical Identifiers

CAS 102-07-8
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00003017
InChI Key GWEHVDNNLFDJLR-UHFFFAOYSA-N
Synonym n,n'-diphenylurea, carbanilide, diphenylurea, diphenylcarbamide, s-diphenylurea, urea, n,n'-diphenyl, sym-diphenylurea, acardite i, acardite, n-phenyl-n'-phenylurea
PubChem CID 7595
ChEBI CHEBI:41320
IUPAC Name 1,3-diphenylurea
SMILES O=C(NC1=CC=CC=C1)NC1=CC=CC=C1

Specifications

Melting Point 240°C
Color White
Boiling Point 260°C
Quantity 25 g
Formula Weight 212.25
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diphenylurea
RTECSNumber FD9800000
TSCA Yes