missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diphenylisobenzofuran 97.0+%, TCI America™
Supplier: TCI America D152025G
Specifications
| 1,3-Diphenylisobenzofuran | |
| 131°C | |
| MFCD00005931 | |
| 1,3-diphenylisobenzofuran, diphenylisobenzofuran, isobenzofuran, 1,3-diphenyl, 1,3 diphenylisobenzofuran, 2,5-diphenyl-3,4-benzofuran, acmc-209lj9, 1,3-diphenylbenzo c furan, 1,3-diphenylisobenzo b furan, 1,3-di phenyl-2-benzofuran, # | |
| C1=CC=C(C=C1)C2=C3C=CC=CC3=C(O2)C4=CC=CC=C4 | |
| 270.331 | |
| 270.33 | |
| Crystalline Powder |
| 5471-63-6 | |
| C20H14O | |
| 25 g | |
| ZKSVYBRJSMBDMV-UHFFFAOYSA-N | |
| 1,3-diphenyl-2-benzofuran | |
| 21649 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 5471-63-6 | |
| 270.331 | |
| ZKSVYBRJSMBDMV-UHFFFAOYSA-N | |
| 21649 | |
| C1=CC=C(C=C1)C2=C3C=CC=CC3=C(O2)C4=CC=CC=C4 |
| C20H14O | |
| MFCD00005931 | |
| 1,3-diphenylisobenzofuran, diphenylisobenzofuran, isobenzofuran, 1,3-diphenyl, 1,3 diphenylisobenzofuran, 2,5-diphenyl-3,4-benzofuran, acmc-209lj9, 1,3-diphenylbenzo c furan, 1,3-diphenylisobenzo b furan, 1,3-di phenyl-2-benzofuran, # | |
| 1,3-diphenyl-2-benzofuran |
Safety and Handling
TSCA : No