missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylisobenzofuran 97.0+%, TCI America™

Catalog No. p-7126763
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15205G 5 g
D152025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15205G Supplier TCI America Supplier No. D15205G
Only null left

Chemical Identifiers

CAS 5471-63-6
Molecular Formula C20H14O
Molecular Weight (g/mol) 270.331
MDL Number MFCD00005931
InChI Key ZKSVYBRJSMBDMV-UHFFFAOYSA-N
Synonym 1,3-diphenylisobenzofuran, diphenylisobenzofuran, isobenzofuran, 1,3-diphenyl, 1,3 diphenylisobenzofuran, 2,5-diphenyl-3,4-benzofuran, acmc-209lj9, 1,3-diphenylbenzo c furan, 1,3-diphenylisobenzo b furan, 1,3-di phenyl-2-benzofuran, #
PubChem CID 21649
IUPAC Name 1,3-diphenyl-2-benzofuran
SMILES C1=CC=C(C=C1)C2=C3C=CC=CC3=C(O2)C4=CC=CC=C4

Specifications

Melting Point 131°C
Quantity 5 g
Formula Weight 270.33
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diphenylisobenzofuran