missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylacetone p-Toluenesulfonylhydrazone 98.0+%, TCI America™

Catalog No. p-7127103
Change view
Click to view available options
Quantity:
10 g
25 g
Catalog No. Quantity
D209310G 10 g
D209325G 25 g
2 options

Catalog No. D209310G

Supplier: TCI America D209310G

Only null left

Chemical Identifiers

CAS 19816-88-7
Molecular Formula C22H22N2O2S
Molecular Weight (g/mol) 378.49
MDL Number MFCD00009645
InChI Key GDXUEUWCUUAZFM-UHFFFAOYSA-N
Synonym 1,3-diphenylacetone p-toluenesulfonylhydrazone, 1,3-diphenylacetone p-tosylhydrazone, 1,3-diphenyl-2-propanone tosylhydrazone, 1,3-diphenylpropan-2-one tosylhydrazone, n'-1,3-diphenylpropan-2-ylidene-4-methylbenzenesulfonohydrazide, 1,3-diphenyl-2-propanone p-tosylhydrazone, n-1,3-diphenylpropan-2-ylideneamino-4-methylbenzenesulfonamide, acmc-209f2n, dibenzyl ketone tosyl hydrazone, benzenesulfonic acid, 4-methyl-, 2-phenyl-1-phenylmethyl ethylidene hydrazide
PubChem CID 88263
IUPAC Name N'-(1,3-diphenylpropan-2-ylidene)-4-methylbenzene-1-sulfonohydrazide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NN=C(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

Melting Point 185°C
Color White-Yellow
Quantity 10 g
Formula Weight 378.49
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diphenylacetone p-Toluenesulfonylhydrazone