missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenyl-2-thiourea 98.0+%, TCI America™

Catalog No. p-7136137
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T019725G 25 g
T0197500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T019725G Supplier TCI America Supplier No. T019725G
Only null left

Chemical Identifiers

CAS 102-08-9
Molecular Formula C13H12N2S
Molecular Weight (g/mol) 228.31
MDL Number MFCD00004921
InChI Key FCSHMCFRCYZTRQ-UHFFFAOYSA-N
Synonym thiocarbanilide, diphenylthiourea, 1,3-diphenyl-2-thiourea, n,n'-diphenylthiourea, sulfocarbanilide, sym-diphenylthiourea, thiourea, n,n'-diphenyl, stabilisator c, rhenocure ca, vulkacit ca
PubChem CID 700999
IUPAC Name 1,3-diphenylthiourea
SMILES S=C(NC1=CC=CC=C1)NC1=CC=CC=C1

Specifications

Melting Point 151°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 228.31
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diphenyl-2-thiourea
EINECSNumber (3)-2205
RTECSNumber FE1225000
TSCA Yes