missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diphenoxybenzene 98.0+%, TCI America™
$142.46 - $648.79
Chemical Identifiers
| CAS | 3379-38-2 |
|---|---|
| Molecular Formula | C18H14O2 |
| Molecular Weight (g/mol) | 262.31 |
| MDL Number | MFCD00003036 |
| InChI Key | JTNRGGLCSLZOOQ-UHFFFAOYSA-N |
| Synonym | m-diphenoxybenzene, benzene, 1,3-diphenoxy, m-diphenyloxybenzene, m-phenoxyphenoxybenzene, benzene, m-diphenoxy, resorcinol diphenyl ether, 1,1'-1,3-phenylenebis oxy dibenzene, m-diphenoxybenzol, acmc-1adxk |
| PubChem CID | 76911 |
| ChEBI | CHEBI:39273 |
| IUPAC Name | 1,3-diphenoxybenzene |
| SMILES | O(C1=CC=CC=C1)C1=CC(OC2=CC=CC=C2)=CC=C1 |
Chemical Identifiers
| 3379-38-2 | |
| 262.31 | |
| JTNRGGLCSLZOOQ-UHFFFAOYSA-N | |
| 76911 | |
| 1,3-diphenoxybenzene |
| C18H14O2 | |
| MFCD00003036 | |
| m-diphenoxybenzene, benzene, 1,3-diphenoxy, m-diphenyloxybenzene, m-phenoxyphenoxybenzene, benzene, m-diphenoxy, resorcinol diphenyl ether, 1,1'-1,3-phenylenebis oxy dibenzene, m-diphenoxybenzol, acmc-1adxk | |
| CHEBI:39273 | |
| O(C1=CC=CC=C1)C1=CC(OC2=CC=CC=C2)=CC=C1 |
Specifications
| 3379-38-2 | |
| White-Yellow | |
| C18H14O2 | |
| 5 g | |
| JTNRGGLCSLZOOQ-UHFFFAOYSA-N | |
| 1,3-diphenoxybenzene | |
| 76911 | |
| 262.31 | |
| Crystalline Powder |
| 60°C | |
| 189°C | |
| MFCD00003036 | |
| m-diphenoxybenzene, benzene, 1,3-diphenoxy, m-diphenyloxybenzene, m-phenoxyphenoxybenzene, benzene, m-diphenoxy, resorcinol diphenyl ether, 1,1'-1,3-phenylenebis oxy dibenzene, m-diphenoxybenzol, acmc-1adxk | |
| O(C1=CC=CC=C1)C1=CC(OC2=CC=CC=C2)=CC=C1 | |
| 262.31 | |
| CHEBI:39273 | |
| ≥98.0% (GC) | |
| 1,3-Diphenoxybenzene |
Safety and Handling
TSCA : Yes