missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenoxybenzene 98.0+%, TCI America™

Catalog No. D26625G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
D26625G 5 g
D266225G 25 g
2 options

Catalog No. D26625G

Supplier: TCI America D26625G

Only null left

Chemical Identifiers

CAS 3379-38-2
Molecular Formula C18H14O2
Molecular Weight (g/mol) 262.31
MDL Number MFCD00003036
InChI Key JTNRGGLCSLZOOQ-UHFFFAOYSA-N
Synonym m-diphenoxybenzene, benzene, 1,3-diphenoxy, m-diphenyloxybenzene, m-phenoxyphenoxybenzene, benzene, m-diphenoxy, resorcinol diphenyl ether, 1,1'-1,3-phenylenebis oxy dibenzene, m-diphenoxybenzol, acmc-1adxk
PubChem CID 76911
ChEBI CHEBI:39273
IUPAC Name 1,3-diphenoxybenzene
SMILES O(C1=CC=CC=C1)C1=CC(OC2=CC=CC=C2)=CC=C1

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 189°C
Quantity 5 g
Formula Weight 262.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diphenoxybenzene