Learn More
(1,3-Dioxolan-2-ylmethyl)magnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific™
Chemical Identifiers
| CAS | 180675-22-3 |
|---|---|
| Molecular Formula | C4H7BrMgO2 |
| Molecular Weight (g/mol) | 191.31 |
| MDL Number | MFCD01321271 |
| InChI Key | IERSPCAGKAOLSQ-UHFFFAOYSA-M |
| Synonym | 1,3-dioxolan-2-ylmethyl magnesium bromide solution, magnesium 2-methanidyl-1,3-dioxolane bromide, magnesium 2+ ion 1,3-dioxolan-2-ylmethanide bromide, grignard reagent |
| PubChem CID | 4247254 |
| IUPAC Name | magnesium;2-methanidyl-1,3-dioxolane;bromide |
| SMILES | [Mg++].[Br-].[CH2-]C1OCCO1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC438861000
|
Thermo Scientific Chemicals
438861000 |
100mL | AcroSeal™ Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC438868000
|
Thermo Scientific Chemicals
438868000 |
800mL | AcroSeal™ Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 180675-22-3 | |
| 191.31 | |
| IERSPCAGKAOLSQ-UHFFFAOYSA-M | |
| 4247254 | |
| [Mg++].[Br-].[CH2-]C1OCCO1 |
| C4H7BrMgO2 | |
| MFCD01321271 | |
| 1,3-dioxolan-2-ylmethyl magnesium bromide solution, magnesium 2-methanidyl-1,3-dioxolane bromide, magnesium 2+ ion 1,3-dioxolan-2-ylmethanide bromide, grignard reagent | |
| magnesium;2-methanidyl-1,3-dioxolane;bromide |
Specifications
| 180675-22-3,109-99-9 | |
| 12.21,91.86 | |
| 65.0°C to 67.0°C | |
| AcroSeal™ Glass Bottle | |
| MFCD01321271 | |
| 0.94 | |
| IERSPCAGKAOLSQ-UHFFFAOYSA-M | |
| magnesium;2-methanidyl-1,3-dioxolane;bromide | |
| 4247254 | |
| (1, 3-Dioxolan-2-ylmethyl)magnesium bromide |
| 8.14,87.79 | |
| 0.9400g/mL | |
| −17°C | |
| C4H7BrMgO2 | |
| 100mL | |
| 1,3-dioxolan-2-ylmethyl magnesium bromide solution, magnesium 2-methanidyl-1,3-dioxolane bromide, magnesium 2+ ion 1,3-dioxolan-2-ylmethanide bromide, grignard reagent | |
| [Mg++].[Br-].[CH2-]C1OCCO1 | |
| 191.31 | |
| 191.31 |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.
May form explosive peroxides.
Suspected of causing cancer.<br/
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
RUO â Research Use Only