missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxane 98.0+%, TCI America™

Catalog No. p-7126470
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D085925ML 25 mL
D0859100ML 100 mL
D0859500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D085925ML Supplier TCI America Supplier No. D085925ML
Only null left

Chemical Identifiers

CAS 505-22-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006566
InChI Key VDFVNEFVBPFDSB-UHFFFAOYSA-N
Synonym m-dioxane, 1,3-dioxacyclohexane, meta-dioxane, 1,3-propanediol formal, m-dioxin, dihydro, trimethylene glycol methylene ether, dihydro-m-dioxin, unii-b2c8m17i09, ccris 5911, 2,4-dioxane
PubChem CID 10450
ChEBI CHEBI:46924
IUPAC Name 1,3-dioxane
SMILES C1COCOC1

Specifications

Color Colorless
Boiling Point 105°C
Quantity 25 mL
UN Number 1165
Formula Weight 88.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dioxane
RTECSNumber JG8224000
TSCA No