accessibility menu, dialog, popup

UserName

CAS RN 505-22-6

IUPAC Name: 1,3-dioxane
Synonyms: 1,3-dioxane trimethylene glycol methylene ether 1,3-propanediol formal dihydro-m-dioxin m-dioxin, dihydro-
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: VDFVNEFVBPFDSB-UHFFFAOYSA-N
SMILES: C1COCOC1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)