missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxane, 98%

Catalog No. p-7035883
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5410706 5 g
AAH5410714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5410706 Supplier Thermo Scientific Chemicals Supplier No. H5410706

CAS: 505-22-6 | C4H8O2 | 88.106 g/mol

1,3-Dioxane has been employed as solvent in the synthesis of 8- and 9-membered dioxazocines and dioxazonines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dioxane has been employed as solvent in the synthesis of 8- and 9-membered dioxazocines and dioxazonines.

Solubility
Fully miscible in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 505-22-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006566
InChI Key VDFVNEFVBPFDSB-UHFFFAOYSA-N
Synonym m-dioxane, 1,3-dioxacyclohexane, meta-dioxane, 1,3-propanediol formal, m-dioxin, dihydro, trimethylene glycol methylene ether, dihydro-m-dioxin, unii-b2c8m17i09, ccris 5911, 2,4-dioxane
PubChem CID 10450
ChEBI CHEBI:46924
IUPAC Name 1,3-dioxane
SMILES C1COCOC1

Specifications

Melting Point -45°C
Density 1.032
Boiling Point 105°C to 106°C
Flash Point 5°C (41°F)
Refractive Index 1.418
Quantity 5 g
UN Number UN1165
Beilstein 102532
Solubility Information Fully miscible in water.
Formula Weight 88.11
Percent Purity 98%
Chemical Name or Material 1,3-Dioxane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIOXANE
EINECSNumber 208-005-1
RTECSNumber JG8224000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only