missing translation for 'onlineSavingsMsg'
Learn More

(1,3-Dioxan-2-ylethyl)magnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. ac431900500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431900500 50 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431900500 Supplier Thermo Scientific Chemicals Supplier No. 431900500
Only null left

CAS: 78078-50-9 | C6H11BrMgO2 | 219.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78078-50-9 , 109-99-9
Molecular Formula C6H11BrMgO2
Molecular Weight (g/mol) 219.36
MDL Number MFCD00061518
InChI Key JYNXRXBIEHSSLR-UHFFFAOYSA-M
Synonym 1,3-dioxan-2-ylethyl magnesium bromide, 1,3-dioxan-2-ylethyl magnesium bromide solution, jynxrxbiehsslr-uhfffaoysa-m, 1,3-dioxan-2-ylethyl magnesiumbromide, 1,3-dioxane-2-ylethyl magnesium bromide, 1,3-dioxan-2-ylethyl-magnesium bromide, 2-2-bromomagnesio ethyl-1,3-dioxane, 2-1,3-dioxan-2-yl ethylmagnesium bromide, 2-1,3-dioxan-2-yl ethyl magnesium bromide, grignard reagent
PubChem CID 11031353
IUPAC Name 2-[2-(bromomagnesio)ethyl]-1,3-dioxane
SMILES Br[Mg]CCC1OCCCO1

Specifications

CAS Min % 9.23, 86.16
CAS Max % 13.84, 90.77
Color White
Density 0.9510g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Quantity 50 mL
Specific Gravity 0.951
Solubility Information Solubility in water: reacts
Formula Weight 219.36
Physical Form Crystalline Powder or Powder
Chemical Name or Material (1, 3-Dioxan-2-ylethyl)magnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only