missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxan-2-one 98.0+%, TCI America™

Catalog No. D38215G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38215G 5 g
D382125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38215G Supplier TCI America Supplier No. D38215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2453-03-4
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00010440
InChI Key YFHICDDUDORKJB-UHFFFAOYSA-N
Synonym Trimethylene Carbonate
PubChem CID 123834
IUPAC Name 1,3-dioxan-2-one
SMILES C1COC(=O)OC1

Specifications

Melting Point 48°C
Color White
Boiling Point 135°C
Quantity 5 g
Formula Weight 102.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dioxan-2-one