accessibility menu, dialog, popup

UserName

CAS RN 2453-03-4

OOO

CAS RN 2453-03-4

IUPAC Name: 1,3-dioxan-2-one
Synonyms: trimethylene carbonate
Molecular Weight (g/mol): 102.09
Molecular Formula: C4H6O3
InChi Key: YFHICDDUDORKJB-UHFFFAOYSA-N
SMILES: O=C1OCCCO1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results