missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dinonadecanoin, ≥99% (GC), MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| CAS | 372490-74-9 |
| Physical Form | Neat |
| Linear Formula | C41H80O5 |
| MDL Number | MFCD00673512 |
| Synonym | 1,3-Dinonadecanoyl-rac-glycerol; 1,3-Dinonadecanoylglycerol; DG(19:0/0:0/19:0) [rac]; Glyceryl dinonadecanoate |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 653.07 |
| Percent Purity | ≥99% (GC) |
| Grade | Analytical Standard |