missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyluracil 98.0+%, TCI America™

Catalog No. D08085G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D08085G 5 g
D08081G 1 g
D080825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D08085G Supplier TCI America Supplier No. D08085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 874-14-6
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00038065
InChI Key JSDBKAHWADVXFU-UHFFFAOYSA-N
Synonym 1,3-dimethyluracil, n1,n3-dimethyluracil, n,n'-dimethyluracil, 1,3-dimethylpyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 1,3-dimethyl, 2,4-dihydroxy-1,3-dimethylpyrimidine, uracil, 1,3-dimethyl, 1,3-dimethyl-2,4 1h,3h-pyrimidinedione, 1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 1,3-dimethyl-1h-pyrimidine-2,4-dione
PubChem CID 70122
ChEBI CHEBI:74763
IUPAC Name 1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CN1C=CC(=O)N(C)C1=O

Specifications

Melting Point 125°C
Color White
Quantity 5 g
Formula Weight 140.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dimethyluracil