accessibility menu, dialog, popup

UserName

CAS RN 874-14-6

IUPAC Name: 1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 1,3-dimethyluracil|N,N'-dimethyluracil|N,n'-dimethyluracil
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: JSDBKAHWADVXFU-UHFFFAOYSA-N
SMILES: CN1C=CC(=O)N(C)C1=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results