missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethylthiourea 97.0+%, TCI America™

Catalog No. D080425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D080425G 25 g
D0804100G 100 g
D0804500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D080425G Supplier TCI America Supplier No. D080425G

Chemical Identifiers

CAS 534-13-4
Molecular Formula C3H8N2S
Molecular Weight (g/mol) 104.171
MDL Number MFCD00004923
InChI Key VLCDUOXHFNUCKK-UHFFFAOYSA-N
Synonym n,n'-dimethylthiourea, 1,3-dimethyl-2-thiourea, thiourea, n,n'-dimethyl, sym-dimethylthiourea, dimethylthiocarbamide, dimethylthiourea, 1,3-dimethylisothiourea, dimethyl-2-thiourea, urea, 1,3-dimethyl-2-thio, unii-8p30pmd17w
PubChem CID 2723631
IUPAC Name 1,3-dimethylthiourea
SMILES CNC(=S)NC

Specifications

Melting Point 65°C
Color White
Quantity 25 g
Formula Weight 104.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dimethylthiourea
EINECSNumber (2)-3266
RTECSNumber YT1520000
TSCA Yes