missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dimethylimidazolium Iodide 98.0+%, TCI America™
$193.86 - $672.92
Chemical Identifiers
| CAS | 4333-62-4 |
|---|---|
| Molecular Formula | C5H9IN2 |
| Molecular Weight (g/mol) | 224.045 |
| MDL Number | MFCD00159685 |
| InChI Key | ARSMIBSHEYKMJT-UHFFFAOYSA-M |
| PubChem CID | 20334 |
| ChEBI | CHEBI:61324 |
| IUPAC Name | 1,3-dimethylimidazol-1-ium;iodide |
| SMILES | CN1C=C[N+](=C1)C.[I-] |
Chemical Identifiers
| 4333-62-4 | |
| 224.045 | |
| ARSMIBSHEYKMJT-UHFFFAOYSA-M | |
| CHEBI:61324 | |
| CN1C=C[N+](=C1)C.[I-] |
| C5H9IN2 | |
| MFCD00159685 | |
| 20334 | |
| 1,3-dimethylimidazol-1-ium;iodide |
Specifications
| 4333-62-4 | |
| White | |
| MFCD00159685 | |
| ARSMIBSHEYKMJT-UHFFFAOYSA-M | |
| 1,3-dimethylimidazol-1-ium;iodide | |
| 20334 | |
| 224.05 | |
| Crystalline Powder |
| 86°C | |
| C5H9IN2 | |
| 5 g | |
| CN1C=C[N+](=C1)C.[I-] | |
| 224.045 | |
| CHEBI:61324 | |
| ≥98.0% (HPLC,T) | |
| 1,3-Dimethylimidazolium Iodide |
Safety and Handling
RTECSNumber : NJ5038500
TSCA : No