accessibility menu, dialog, popup

UserName

CAS RN 4333-62-4

ICH3NNH3C

CAS RN 4333-62-4

IUPAC Name: 1,3-dimethyl-1H-imidazol-3-ium iodide
Synonyms: 1,3-dimethylimidazolium iodide 1-methylimidazole methiodide
Molecular Weight (g/mol): 224.05
Molecular Formula: C5H9IN2
InChi Key: ARSMIBSHEYKMJT-UHFFFAOYSA-M
SMILES: [I-].CN1C=C[N+](C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results