missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone 98.0+%, TCI America™

Catalog No. D2014500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2014500G 500 g
D201425G 25 g
D2014100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D2014500G Supplier TCI America Supplier No. D2014500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7226-23-5
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.175
MDL Number MFCD00006550
InChI Key GUVUOGQBMYCBQP-UHFFFAOYSA-N
Synonym 1,3-dimethyl-3,4,5,6-tetrahydro-2 1h-pyrimidinone, 1,3-dimethyltetrahydropyrimidin-2 1h-one, dmpu, dimethylpropyleneurea, 2 1h-pyrimidinone, tetrahydro-1,3-dimethyl, n,n'-dimethylpropyleneurea, n,n'-dimethyltrimethyleneurea, ccris 4322, n,n'-dimethylpropylene urea, tetrahydro-1,3-dimethyl-2 1h pyrimidine
PubChem CID 81646
IUPAC Name 1,3-dimethyl-1,3-diazinan-2-one
SMILES CN1CCCN(C1=O)C

Specifications

Color Yellow
pH 11 (100g/L H2O soln. 20°C)
Boiling Point 93°C
Quantity 500 g
Formula Weight 128.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone
EINECSNumber (5)-6728
RTECSNumber UW7582500
TSCA Yes