accessibility menu, dialog, popup

UserName

CAS RN 7226-23-5

IUPAC Name: 1,3-dimethyl-1,3-diazinan-2-one
Synonyms: DMPU dimethyl propyleneurea
Molecular Weight (g/mol): 128.18
Molecular Formula: C6H12N2O
InChi Key: GUVUOGQBMYCBQP-UHFFFAOYSA-N
SMILES: CN1CCCN(C)C1=O
Boiling Point 
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)
  • (2)