missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone, 97%, pure

Catalog No. p-120241
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC224641000 100 mL Glass bottle
AC224645000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC224641000 Supplier Thermo Scientific Chemicals Supplier No. 224641000

CAS: 7226-23-5 | C6H12N2O | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 7226-23-5
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.17
MDL Number MFCD00006550
InChI Key GUVUOGQBMYCBQP-UHFFFAOYSA-N
Synonym 1,3-dimethyl-3,4,5,6-tetrahydro-2 1h-pyrimidinone, 1,3-dimethyltetrahydropyrimidin-2 1h-one, dmpu, dimethylpropyleneurea, 2 1h-pyrimidinone, tetrahydro-1,3-dimethyl, n,n'-dimethylpropyleneurea, n,n'-dimethyltrimethyleneurea, ccris 4322, n,n'-dimethylpropylene urea, tetrahydro-1,3-dimethyl-2 1h pyrimidine
PubChem CID 81646
IUPAC Name 1,3-dimethyl-1,3-diazinan-2-one
SMILES CN1CCCN(C1=O)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -20°C
Color Colorless to Yellow
Density 1.0640g/mL
Boiling Point 246°C
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4875 to 1.4895
Quantity 100 mL
Beilstein 24, IV, 32
Fieser 11,207; 13,122; 16,146
Specific Gravity 1.064
Solubility Information Solubility in water: freely soluble
Viscosity 3.41 mPa.s (20°C)
Formula Weight 128.17
Percent Purity 97%
Grade Pure
Physical Form Liquid
Chemical Name or Material 1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Suspected of damaging fertility.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
EINECSNumber 230-625-6
RTECSNumber UW7582500
TSCA TSCA

RUO – Research Use Only