missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dimethoxybenzene 99.0+%, TCI America™
Supplier: TCI America D0628100G
Specifications
| 1,3-Dimethoxybenzene | |
| Yellow | |
| C8H10O2 | |
| 100 g | |
| DPZNOMCNRMUKPS-UHFFFAOYSA-N | |
| 1,3-dimethoxybenzene | |
| 9025 | |
| ≥99.0% (GC) |
| 151-10-0 | |
| 217°C | |
| MFCD00008384 | |
| m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu | |
| COC1=CC(OC)=CC=C1 | |
| 138.17 | |
| 138.17 | |
| Liquid |
Chemical Identifiers
| 151-10-0 | |
| 138.17 | |
| DPZNOMCNRMUKPS-UHFFFAOYSA-N | |
| 9025 | |
| COC1=CC(OC)=CC=C1 |
| C8H10O2 | |
| MFCD00008384 | |
| m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu | |
| 1,3-dimethoxybenzene |
Safety and Handling
EINECSNumber : (3)-0582
RTECSNumber : CZ6474000
TSCA : Yes