missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethoxybenzene 99.0+%, TCI America™

Catalog No. D062825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D062825G 25 g
D0628100G 100 g
D0628500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D062825G Supplier TCI America Supplier No. D062825G

Chemical Identifiers

CAS 151-10-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00008384
InChI Key DPZNOMCNRMUKPS-UHFFFAOYSA-N
Synonym m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu
PubChem CID 9025
IUPAC Name 1,3-dimethoxybenzene
SMILES COC1=CC(OC)=CC=C1

Specifications

Color Yellow
Boiling Point 217°C
Quantity 25 g
Formula Weight 138.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dimethoxybenzene
EINECSNumber (3)-0582
RTECSNumber CZ6474000
TSCA Yes