missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethoxybenzene, 99%

CAS: 151-10-0 | C8H10O2 | 138.17 g/mol

$295.32 - $295.32

Chemical Identifiers

CAS 151-10-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00008384
InChI Key DPZNOMCNRMUKPS-UHFFFAOYSA-N
Synonym m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu
PubChem CID 9025
IUPAC Name 1,3-dimethoxybenzene
SMILES COC1=CC(OC)=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC155235000
SDS View Documents
Thermo Scientific Chemicals
155235000
500 g Glass bottle
Each for $295.32
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

151-10-0
138.17
DPZNOMCNRMUKPS-UHFFFAOYSA-N
9025
COC1=CC(OC)=CC=C1
C8H10O2
MFCD00008384
m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu
1,3-dimethoxybenzene

Specifications

151-10-0
100.0
1.0500g/mL
87°C
98.5% min. (GC)
C8H10O2
C6H4(OCH3)2
500 g
1.05
Solubility in water: immiscible
COC1=CC(OC)=CC=C1
138.17
138.17
Liquid
98.5
Colorless to Brown
85.0°C to 87.0°C (7.0 mmHg)
Authentic
Glass bottle
1.5230 to 1.5250
MFCD00008384
06, 813
m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu
DPZNOMCNRMUKPS-UHFFFAOYSA-N
1,3-dimethoxybenzene
9025
99%
1, 3-Dimethoxybenzene, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 205-783-4

RTECSNumber : CZ6474000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only