Learn More
1,3-Dimethoxybenzene, 98%
CAS: 151-10-0 | C8H10O2 | 138.17 g/mol
$62.32 - $213.60
Chemical Identifiers
| CAS | 151-10-0 |
|---|---|
| Molecular Formula | C8H10O2 |
| Molecular Weight (g/mol) | 138.17 |
| MDL Number | MFCD00008384 |
| InChI Key | DPZNOMCNRMUKPS-UHFFFAOYSA-N |
| Synonym | m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu |
| PubChem CID | 9025 |
| IUPAC Name | 1,3-dimethoxybenzene |
| SMILES | COC1=CC(OC)=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1338022
|
Thermo Scientific Chemicals
A1338022 |
100 g |
Each for $62.32
|
|
|||||
|
AAA1338036
|
Thermo Scientific Chemicals
A1338036 |
500 g |
Each for $213.60
|
|
|||||
Description
1,3-Dimethoxybenzene is used for the preparation of oxathiane spiroketal donors. It is also used for the formation of pi- and O-ylidic complexes with dichlorocarbene. Further, it acts as a flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Dimethoxybenzene is used for the preparation of oxathiane spiroketal donors. It is also used for the formation of pi- and O-ylidic complexes with dichlorocarbene. Further, it acts as a flavoring agent.
Solubility
Miscible with toluene.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 151-10-0 | |
| 138.17 | |
| DPZNOMCNRMUKPS-UHFFFAOYSA-N | |
| 9025 | |
| COC1=CC(OC)=CC=C1 |
| C8H10O2 | |
| MFCD00008384 | |
| m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu | |
| 1,3-dimethoxybenzene |
Specifications
| 151-10-0 | |
| 1.065 | |
| 87°C (188°F) | |
| C8H10O2 | |
| MFCD00008384 | |
| 878582 | |
| Miscible with toluene. | |
| COC1=CC(OC)=CC=C1 | |
| 138.17 | |
| 138.17 | |
| 1,3-Dimethoxybenzene |
| -52°C | |
| 217°C to 218°C | |
| Characteristic | |
| 1.525 | |
| 100 g | |
| m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu | |
| DPZNOMCNRMUKPS-UHFFFAOYSA-N | |
| 1,3-dimethoxybenzene | |
| 9025 | |
| 98% |
Safety and Handling
GHS H Statement
H312-H227
Harmful in contact with skin.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 205-783-4
RTECSNumber : CZ6474000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only