Learn More
1,3-Dimethoxybenzene, 98%
CAS: 151-10-0 | C8H10O2 | 138.17 g/mol
Supplier: Thermo Scientific Chemicals A1338036
Description
1,3-Dimethoxybenzene is used for the preparation of oxathiane spiroketal donors. It is also used for the formation of pi- and O-ylidic complexes with dichlorocarbene. Further, it acts as a flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Dimethoxybenzene is used for the preparation of oxathiane spiroketal donors. It is also used for the formation of pi- and O-ylidic complexes with dichlorocarbene. Further, it acts as a flavoring agent.
Solubility
Miscible with toluene.
Notes
Incompatible with strong oxidizing agents.
Specifications
| 1,3-Dimethoxybenzene | |
| -52°C | |
| 217°C to 218°C | |
| Characteristic | |
| 1.525 | |
| 500 g | |
| m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu | |
| DPZNOMCNRMUKPS-UHFFFAOYSA-N | |
| 1,3-dimethoxybenzene | |
| 9025 | |
| 98% |
| 151-10-0 | |
| 1.065 | |
| 87°C (188°F) | |
| C8H10O2 | |
| MFCD00008384 | |
| 878582 | |
| Miscible with toluene. | |
| COC1=CC(OC)=CC=C1 | |
| 138.17 | |
| 138.17 |
Chemical Identifiers
| 151-10-0 | |
| 138.17 | |
| DPZNOMCNRMUKPS-UHFFFAOYSA-N | |
| 9025 | |
| COC1=CC(OC)=CC=C1 |
| C8H10O2 | |
| MFCD00008384 | |
| m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu | |
| 1,3-dimethoxybenzene |
Safety and Handling
GHS H Statement
H312-H227
Harmful in contact with skin.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 205-783-4
RTECSNumber : CZ6474000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only