missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethoxybenzene, 98%

Catalog No. AAA1338022
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1338022 100 g
AAA1338036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1338022 Supplier Thermo Scientific Chemicals Supplier No. A1338022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 151-10-0 | C8H10O2 | 138.17 g/mol

1,3-Dimethoxybenzene is used for the preparation of oxathiane spiroketal donors. It is also used for the formation of pi- and O-ylidic complexes with dichlorocarbene. Further, it acts as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dimethoxybenzene is used for the preparation of oxathiane spiroketal donors. It is also used for the formation of pi- and O-ylidic complexes with dichlorocarbene. Further, it acts as a flavoring agent.

Solubility
Miscible with toluene.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 151-10-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00008384
InChI Key DPZNOMCNRMUKPS-UHFFFAOYSA-N
Synonym m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu
PubChem CID 9025
IUPAC Name 1,3-dimethoxybenzene
SMILES COC1=CC(OC)=CC=C1

Specifications

Melting Point -52°C
Density 1.065
Boiling Point 217°C to 218°C
Flash Point 87°C (188°F)
Odor Characteristic
Refractive Index 1.525
Quantity 100 g
Beilstein 878582
Solubility Information Miscible with toluene.
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 1,3-Dimethoxybenzene
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H312-H227
Harmful in contact with skin.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 205-783-4
RTECSNumber CZ6474000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only