missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diisopropylimidazolinium Tetrafluoroborate 98.0+%, TCI America™
$445.51 - $1532.25
Chemical Identifiers
| CAS | 137581-18-1 |
|---|---|
| Molecular Formula | C9H19BF4N2 |
| MDL Number | MFCD08276803 |
| Synonym | IiPrim.HBF4 |
Chemical Identifiers
| 137581-18-1 | |
| C9H19BF4N2 | |
| MFCD08276803 | |
| IiPrim.HBF4 |
Specifications
| 137581-18-1 | |
| White-Yellow | |
| MFCD08276803 | |
| 1759 | |
| 242.07 | |
| Crystalline Powder |
| 146°C | |
| C9H19BF4N2 | |
| 1 g | |
| IiPrim.HBF4 | |
| ≥98.0% (HPLC,N) | |
| 1,3-Diisopropylimidazolinium Tetrafluoroborate |
Safety and Handling
TSCA : No