missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diisopropylimidazolinium Tetrafluoroborate 98.0+%, TCI America™
Supplier: TCI America D36225G
Specifications
| 1,3-Diisopropylimidazolinium Tetrafluoroborate | |
| 146°C | |
| C9H19BF4N2 | |
| 5 g | |
| IiPrim.HBF4 | |
| ≥98.0% (HPLC,N) |
| 137581-18-1 | |
| White-Yellow | |
| MFCD08276803 | |
| 1759 | |
| 242.07 | |
| Crystalline Powder |
Chemical Identifiers
| 137581-18-1 | |
| C9H19BF4N2 | |
| MFCD08276803 | |
| IiPrim.HBF4 |
Safety and Handling
TSCA : No