missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diisopropylbenzene 96.0+%, TCI America™

Catalog No. D249025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D249025ML 25 mL
D2490500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D249025ML Supplier TCI America Supplier No. D249025ML

Chemical Identifiers

CAS 99-62-7
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00008889
InChI Key UNEATYXSUBPPKP-UHFFFAOYSA-N
Synonym 1,3-diisopropylbenzene, m-diisopropylbenzene, benzene, 1,3-bis 1-methylethyl, benzene, m-diisopropyl, m-diisopropylbenzol, 1,3-bis 1-methylethyl benzene, 3-isopropylcumene, unii-482vxw192e, meta-diisopropylbenzene, chembl31352
PubChem CID 7450
IUPAC Name 1,3-di(propan-2-yl)benzene
SMILES CC(C)C1=CC(=CC=C1)C(C)C

Specifications

Color Colorless
Boiling Point 204°C
Quantity 25 mL
UN Number 3082
Formula Weight 162.28
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Diisopropylbenzene
EINECSNumber (3)-0025
RTECSNumber CZ6334000
TSCA Yes