missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diisopropylbenzene, 96%

Catalog No. p-7027275
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2390418 50 g
AAB2390430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2390418 Supplier Thermo Scientific Chemicals Supplier No. B2390418

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-62-7 | C12H18 | 162.276 g/mol

1,3-Diisopropylbenzene was used in the synthesis of benzene-1,3,5-triphosphonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diisopropylbenzene was used in the synthesis of benzene-1,3,5-triphosphonic acid.

Solubility
Soluble in water 4.33 mg/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.

Chemical Identifiers

CAS 99-62-7
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00008889
InChI Key UNEATYXSUBPPKP-UHFFFAOYSA-N
Synonym 1,3-diisopropylbenzene, m-diisopropylbenzene, benzene, 1,3-bis 1-methylethyl, benzene, m-diisopropyl, m-diisopropylbenzol, 1,3-bis 1-methylethyl benzene, 3-isopropylcumene, unii-482vxw192e, meta-diisopropylbenzene, chembl31352
PubChem CID 7450
IUPAC Name 1,3-di(propan-2-yl)benzene
SMILES CC(C)C1=CC(=CC=C1)C(C)C

Specifications

Melting Point -63°C
Density 0.856
Boiling Point 204°C
Flash Point 76°C (170°F)
Refractive Index 1.489
Quantity 50 g
Beilstein 1905828
Solubility Information Soluble in water 4.33mg/L at 25°C.
Formula Weight 162.27
Percent Purity 96%
Chemical Name or Material 1,3-Diisopropylbenzene
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 202-773-1
RTECSNumber CZ6334000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only