missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diiodo-5,5-dimethylhydantoin 97.0+%, TCI America™

Catalog No. D36575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36575G 5 g
D365725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36575G Supplier TCI America Supplier No. D36575G

Chemical Identifiers

CAS 2232-12-4
Molecular Formula C5H6I2N2O2
Molecular Weight (g/mol) 379.92
MDL Number MFCD00020867
InChI Key RDZHCKRAHUPIFK-UHFFFAOYSA-N
Synonym DIH
PubChem CID 200534
IUPAC Name 1,3-diiodo-5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C(=O)N(C(=O)N1I)I)C

Specifications

Melting Point 198°C
Quantity 5 g
UN Number 3085
Formula Weight 379.92
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diiodo-5,5-dimethylhydantoin
EINECSNumber (5)-7009
RTECSNumber MU0970000
TSCA No
Recommended Storage Freezer