accessibility menu, dialog, popup

UserName

CH3H3CNIONIO

CAS RN 2232-12-4

IUPAC Name: 1,3-diiodo-5,5-dimethylimidazolidine-2,4-dione
Synonyms: 1,3-diiodo-5,5-dimethylimidazolidine-2,4-dione
Molecular Weight (g/mol): 379.92
Molecular Formula: C5H6I2N2O2
InChi Key: RDZHCKRAHUPIFK-UHFFFAOYSA-N
SMILES: CC1(C)N(I)C(=O)N(I)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results