missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diiminoisoindoline 98.0+%, TCI America™
$71.23 - $215.62
Chemical Identifiers
| CAS | 57500-34-2 |
|---|---|
| Molecular Formula | C8H9N3 |
| Molecular Weight (g/mol) | 147.18 |
| MDL Number | MFCD00010062 |
| InChI Key | RZVCEPSDYHAHLX-UHFFFAOYSA-P |
| PubChem CID | 18980 |
| IUPAC Name | 3-amino-1-iminiumyl-1H-isoindol-2-ium |
| SMILES | NC1=[NH+]C(=[NH2+])C2=CC=CC=C12 |
Chemical Identifiers
| 57500-34-2 | |
| 147.18 | |
| RZVCEPSDYHAHLX-UHFFFAOYSA-P | |
| 3-amino-1-iminiumyl-1H-isoindol-2-ium |
| C8H9N3 | |
| MFCD00010062 | |
| 18980 | |
| NC1=[NH+]C(=[NH2+])C2=CC=CC=C12 |
Specifications
| 57500-34-2 | |
| C8H9N3 | |
| 5 g | |
| NC1=[NH+]C(=[NH2+])C2=CC=CC=C12 | |
| 147.18 | |
| 145.17 | |
| Crystalline Powder |
| 195°C | |
| MFCD00010062 | |
| RZVCEPSDYHAHLX-UHFFFAOYSA-P | |
| 3-amino-1-iminiumyl-1H-isoindol-2-ium | |
| 18980 | |
| ≥98.0% (T) | |
| 1,3-Diiminoisoindoline |
Safety and Handling
RTECSNumber : NR3500000
TSCA : No