CAS RN 57500-34-2
CAS RN 57500-34-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,3-Diiminoisoindoline 98.0+%, TCI America™
CAS: 57500-34-2 Molecular Formula: C8H9N3 Molecular Weight (g/mol): 147.18 MDL Number: MFCD00010062 InChI Key: RZVCEPSDYHAHLX-UHFFFAOYSA-P PubChem CID: 18980 IUPAC Name: 3-amino-1-iminiumyl-1H-isoindol-2-ium SMILES: NC1=[NH+]C(=[NH2+])C2=CC=CC=C12