missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diiminoisoindoline 98.0+%, TCI America™

Catalog No. D20795G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20795G 5 g
D207925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20795G Supplier TCI America Supplier No. D20795G

Chemical Identifiers

CAS 57500-34-2
Molecular Formula C8H9N3
Molecular Weight (g/mol) 147.18
MDL Number MFCD00010062
InChI Key RZVCEPSDYHAHLX-UHFFFAOYSA-P
PubChem CID 18980
IUPAC Name 3-amino-1-iminiumyl-1H-isoindol-2-ium
SMILES NC1=[NH+]C(=[NH2+])C2=CC=CC=C12

Specifications

Melting Point 195°C
Quantity 5 g
Formula Weight 145.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diiminoisoindoline
RTECSNumber NR3500000
TSCA No