missing translation for 'onlineSavingsMsg'
Learn More

1,3-Difluoro-5-iodobenzene 98.0+%, TCI America™

Catalog No. D46205G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46205G 5 g
D462025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46205G Supplier TCI America Supplier No. D46205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2265-91-0
Molecular Formula C6H3F2I
Molecular Weight (g/mol) 239.99
MDL Number MFCD03094155
InChI Key QQCFOQNFPIAENW-UHFFFAOYSA-N
Synonym 3,5-difluoroiodobenzene, benzene, 1,3-difluoro-5-iodo, 1,3-difluoro-5-iodo-benzene, 3,5-difluoro iodobenzene, pubchem17134, 3,5-difluoro-iodobenzene, 3-difluoro-5-iodobenzene
PubChem CID 2778221
IUPAC Name 1,3-difluoro-5-iodobenzene
SMILES FC1=CC(I)=CC(F)=C1

Specifications

Color Yellow
Boiling Point 60°C
Quantity 5 g
Formula Weight 239.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Difluoro-5-iodobenzene