missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethylurea 98.0+%, TCI America™

Catalog No. D053425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D053425G 25 g
D0534500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D053425G Supplier TCI America Supplier No. D053425G

Chemical Identifiers

CAS 623-76-7
Molecular Formula C5H12N2O
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009028
InChI Key ZWAVGZYKJNOTPX-UHFFFAOYSA-N
Synonym n,n'-diethylurea, sym-diethylurea, urea, n,n'-diethyl, urea, 1,3-diethyl, urea, n,n'-diethyl 9ci, sym-n,n'-diethylurea, n-ethyl ethylamino carboxamide, urea,3-diethyl, urea,n'-diethyl, 1,3-diethyl-urea
PubChem CID 12194
IUPAC Name 1,3-diethylurea
SMILES CCNC(=O)NCC

Specifications

Melting Point 111°C
Color White
Quantity 25 g
Formula Weight 116.16
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diethylurea
RTECSNumber YS9354000
TSCA No